Assay Design



Gene Transcripts

Gene Symbol
Cry1
Entrez Gene ID
299691
Gene Name
cryptochrome circadian clock 1
Gene Aliases
-
Chromosome Location
Chr.7: 24534591 - 24634098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198750.2 NP_942045.2 11 - 12 2203 92
XM_008765225.1 XP_008763447.1 11 - 12 2227 92
GenBank mRNA AF545855.1 - 11 - 12 1767 92
BC107677.1 - 11 - 12 2203 92

View Full Product Details