Assay Design



Gene Transcripts

Gene Symbol
Ccdc38
Entrez Gene ID
500823
Gene Name
coiled-coil domain containing 38
Gene Aliases
RGD1564046
Chromosome Location
Chr.7: 34402930 - 34477861 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241271.2 XP_006241333.1 16 - 17 1731 96
XM_008765310.1 XP_008763532.1 15 - 16 1461 96

View Full Product Details