Assay Design



Gene Transcripts

Gene Symbol
Frs2
Entrez Gene ID
314850
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
-
Chromosome Location
Chr.7: 60128500 - 60210499 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108097.3 NP_001101567.2 7 - 8 700 60
XM_006241381.2 XP_006241443.1 5 - 6 642 60
GenBank mRNA CB581691.1 - 5 - 6 523 60

View Full Product Details