Assay Design



Gene Transcripts

Gene Symbol
Nfam1
Entrez Gene ID
362966
Gene Name
NFAT activating protein with ITAM motif 1
Gene Aliases
RGD1308316
Chromosome Location
Chr.7: 123856718 - 123893792 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242140.2 XP_006242202.1 5 - 6 876 75
XM_006242142.2 XP_006242204.1 5 - 6 789 75

View Full Product Details