Assay Design



Gene Transcripts

Gene Symbol
Mal2
Entrez Gene ID
362911
Gene Name
mal, T-cell differentiation protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 94130852 - 94163649 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198786.2 NP_942081.2 3 - 4 615 89
GenBank mRNA AY079080.1 - 3 - 4 475 89
AY079081.2 - 2 - 3 728 89
BC062388.1 - 3 - 4 615 89

View Full Product Details