Assay Design



Gene Transcripts

Gene Symbol
Poldip3
Entrez Gene ID
315170
Gene Name
polymerase (DNA-directed), delta interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 123992167 - 124020596 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130506.1 NP_001123978.1 6 - 7 989 64
XM_006242090.2 XP_006242152.1 5 - 6 924 64

View Full Product Details