Assay Design



Gene Transcripts

Gene Symbol
Mbd6
Entrez Gene ID
362892
Gene Name
methyl-CpG binding domain protein 6
Gene Aliases
-
Chromosome Location
Chr.7: 70571435 - 70580163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241463.2 XP_006241525.1 6 - 7 1822 88
XM_008765404.1 XP_008763626.1 5 - 6 1513 88
XM_008765405.1 XP_008763627.1 6 - 7 1817 88
XM_008765406.1 XP_008763628.1 6 - 7 1652 88
XM_008765407.1 XP_008763629.1 5 - 6 1513 88

View Full Product Details