Assay Design



Gene Transcripts

Gene Symbol
Dtx3
Entrez Gene ID
500847
Gene Name
deltex 3, E3 ubiquitin ligase
Gene Aliases
RGD1566181
Chromosome Location
Chr.7: 70476737 - 70482853 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191989.1 NP_001178918.1 5 - 6 1287 90
XM_006241495.2 XP_006241557.1 5 - 6 2322 90

View Full Product Details