Assay Design



Gene Transcripts

Gene Symbol
Josd1
Entrez Gene ID
315134
Gene Name
Josephin domain containing 1
Gene Aliases
RGD1305388
Chromosome Location
Chr.7: 120905343 - 120919261 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025009.1 NP_001020180.1 3 - 4 707 114
GenBank mRNA BC091280.1 - 3 - 4 707 114

View Full Product Details