Assay Design



Gene Transcripts

Gene Symbol
Ncf4
Entrez Gene ID
500904
Gene Name
neutrophil cytosolic factor 4
Gene Aliases
-
Chromosome Location
Chr.7: 119481708 - 119499426 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127304.1 NP_001120776.1 8 - 9 989 102
XM_006241939.2 XP_006242001.1 9 - 10 1653 102
GenBank mRNA BC167076.1 - 8 - 9 961 102

View Full Product Details