Assay Design
Gene Transcripts
- Gene Symbol
- Ago2
- Entrez Gene ID
-
59117
- Gene Name
- argonaute RISC catalytic component 2
- Gene Aliases
-
Eif2c2
- Chromosome Location
-
Chr.7: 114339607 - 114377277 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_021597.1 |
NP_067608.1 |
2 - 3 |
347 |
119 |
GenBank mRNA |
AF195534.1 |
- |
2 - 3 |
347 |
119 |
View Full Product Details