Assay Design



Gene Transcripts

Gene Symbol
Oplah
Entrez Gene ID
116684
Gene Name
5-oxoprolinase (ATP-hydrolysing)
Gene Aliases
-
Chromosome Location
Chr.7: 117353954 - 117369730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053904.1 NP_446356.1 7 - 8 1057 89
XM_006241771.2 XP_006241833.1 7 - 8 2717 89
XM_006241772.2 XP_006241834.1 7 - 8 2717 89
XM_006241773.2 XP_006241835.1 7 - 8 2600 89
XM_006241774.2 XP_006241836.1 7 - 8 1395 89
XM_006241775.2 XP_006241837.1 8 - 9 1254 89
GenBank mRNA BC085330.1 - 7 - 8 1122 89
U70825.1 - 7 - 8 1057 89

View Full Product Details