Assay Design



Gene Transcripts

Gene Symbol
Nrbp2
Entrez Gene ID
680451
Gene Name
nuclear receptor binding protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 117145106 - 117151694 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135007.1 NP_001128479.1 16 - 17 1873 67
XM_006241885.2 XP_006241947.1 17 - 18 1892 67
GenBank mRNA FQ223130.1 - 11 - 12 1434 67

View Full Product Details