Assay Design



Gene Transcripts

Gene Symbol
Lrrc14
Entrez Gene ID
500900
Gene Name
leucine rich repeat containing 14
Gene Aliases
-
Chromosome Location
Chr.7: 117773218 - 117779520 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024354.1 NP_001019525.1 1 - 2 149 93
GenBank mRNA BC092583.1 - 1 - 2 149 93

View Full Product Details