Assay Design



Gene Transcripts

Gene Symbol
Shisa8
Entrez Gene ID
315163
Gene Name
shisa family member 8
Gene Aliases
RGD1563996
Chromosome Location
Chr.7: 123440163 - 123445613 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207022.1 NP_001193951.1 1 - 2 1023 82

View Full Product Details