Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144748.1 | NP_653349.1 | 11 - 12 | 1512 | 118 |
XM_008759677.1 | XP_008757899.1 | 11 - 12 | 1497 | 118 | |
XM_008759678.1 | XP_008757900.1 | 10 - 11 | 1453 | 118 | |
XM_008759679.1 | XP_008757901.1 | 8 - 9 | 1250 | 118 | |
GenBank mRNA | AF062389.1 | - | 11 - 12 | 1512 | 118 |
BC078721.1 | - | 11 - 12 | 1489 | 118 |