Assay Design



Gene Transcripts

Gene Symbol
Kcne3
Entrez Gene ID
63883
Gene Name
potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 3
Gene Aliases
-
Chromosome Location
Chr.1: 165189280 - 165196949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229785.2 XP_006229847.1 2 - 3 2044 132
XM_006229786.2 XP_006229848.1 2 - 3 899 132
GenBank mRNA AJ271742.1 - 2 - 3 187 132

View Full Product Details