Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012345.1 | NP_001012345.1 | 4 - 5 | 613 | 62 |
XM_006229733.2 | XP_006229795.1 | 3 - 4 | 653 | 62 | |
XM_008759689.1 | XP_008757911.1 | 4 - 5 | 364 | 62 | |
GenBank mRNA | BC089846.1 | - | 4 - 5 | 613 | 62 |