Assay Design



Gene Transcripts

Gene Symbol
Acsm1
Entrez Gene ID
361638
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
Bucs1
Chromosome Location
Chr.1: 189359378 - 189395277 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108502.2 NP_001101972.2 3 - 4 514 62
XM_006230127.2 XP_006230189.1 4 - 5 667 62
XM_006230128.2 XP_006230190.1 4 - 5 664 62
XM_006230129.2 XP_006230191.1 3 - 4 430 62
XM_008759771.1 XP_008757993.1 4 - 5 583 62
XM_008759772.1 XP_008757994.1 3 - 4 559 62
XM_008759773.1 XP_008757995.1 4 - 5 664 62
GenBank mRNA BC168985.1 - 3 - 4 514 62

View Full Product Details