Assay Design



Gene Transcripts

Gene Symbol
Qprt
Entrez Gene ID
293504
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
-
Chromosome Location
Chr.1: 198544262 - 198559556 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009646.1 NP_001009646.1 3 - 4 731 159
XM_006230246.2 XP_006230308.1 3 - 4 902 159
XM_008759877.1 XP_008758099.1 3 - 4 1207 159
XM_008759878.1 XP_008758100.1 4 - 5 994 159
GenBank mRNA BC088177.1 - 3 - 4 731 159
FQ209866.1 - 3 - 4 725 159

View Full Product Details