Assay Design



Gene Transcripts

Gene Symbol
Il18bp
Entrez Gene ID
84388
Gene Name
interleukin 18 binding protein
Gene Aliases
Igifbp
Chromosome Location
Chr.1: 167091521 - 167095718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053374.2 NP_445826.2 2 - 3 230 100
XM_008759805.1 XP_008758027.1 2 - 3 2388 100
GenBank mRNA CB809932.1 - 2 - 3 241 100

View Full Product Details