Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017082.1 | NP_058778.1 | 8 - 9 | 1862 | 60 |
XM_006230107.2 | XP_006230169.1 | 9 - 10 | 1984 | 60 | |
GenBank mRNA | BC081814.1 | - | 8 - 9 | 1868 | 60 |
M63510.1 | - | 8 - 9 | 1862 | 60 | |
S75960.1 | - | 8 - 9 | 1853 | 60 |