Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017082.1 | NP_058778.1 | 6 - 7 | 1612 | 103 |
XM_006230107.2 | XP_006230169.1 | 7 - 8 | 1734 | 103 | |
GenBank mRNA | BC081814.1 | - | 6 - 7 | 1618 | 103 |
M63510.1 | - | 6 - 7 | 1612 | 103 | |
S75960.1 | - | 6 - 7 | 1603 | 103 |