Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108497.1 | NP_001101967.1 | 1 - 2 | 439 | 102 |
XM_006229987.1 | XP_006230049.1 | 2 - 3 | 328 | 102 | |
XM_006229988.2 | XP_006230050.1 | 2 - 3 | 259 | 102 | |
XM_006229990.1 | XP_006230052.1 | 2 - 3 | 328 | 102 | |
XM_006229992.1 | XP_006230054.1 | 2 - 3 | 328 | 102 |