Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108497.1 | NP_001101967.1 | 4 - 5 | 760 | 111 |
XM_006229987.1 | XP_006230049.1 | 5 - 6 | 649 | 111 | |
XM_006229988.2 | XP_006230050.1 | 5 - 6 | 580 | 111 | |
XM_006229989.1 | XP_006230051.1 | 5 - 6 | 619 | 111 | |
XM_006229990.1 | XP_006230052.1 | 5 - 6 | 607 | 111 | |
XM_006229991.1 | XP_006230053.1 | 4 - 5 | 568 | 111 | |
XM_006229992.1 | XP_006230054.1 | 4 - 5 | 493 | 111 |