Assay Design



Gene Transcripts

Gene Symbol
Kctd21
Entrez Gene ID
499209
Gene Name
potassium channel tetramerization domain containing 21
Gene Aliases
RGD1559529
Chromosome Location
Chr.1: 162307775 - 162325269 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109151.1 NP_001102621.1 1 - 1 559 85
XM_006229781.2 XP_006229843.1 2 - 2 630 85

View Full Product Details