Assay Design



Gene Transcripts

Gene Symbol
Art1
Entrez Gene ID
308873
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.1: 167202064 - 167206615 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107541.2 NP_001101011.1 2 - 3 878 111

View Full Product Details