Assay Design
Gene Transcripts
- Gene Symbol
- Gab2
- Entrez Gene ID
-
84477
- Gene Name
- GRB2-associated binding protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 162083038 - 162282707 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053417.1 |
NP_445869.1 |
1 - 2 |
82 |
65 |
GenBank mRNA |
AF230367.2 |
- |
1 - 2 |
82 |
65 |
View Full Product Details