Assay Design
Gene Transcripts
- Gene Symbol
- Wif1
- Entrez Gene ID
-
114557
- Gene Name
- Wnt inhibitory factor 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 63094932 - 63165158 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053738.1 |
NP_446190.1 |
8 - 9 |
1201 |
101 |
GenBank mRNA |
AY030278.1 |
- |
8 - 9 |
1201 |
101 |
View Full Product Details