Assay Design



Gene Transcripts

Gene Symbol
Llph
Entrez Gene ID
299818
Gene Name
LLP homolog, long-term synaptic facilitation (Aplysia)
Gene Aliases
RGD1308261
Chromosome Location
Chr.7: 65004548 - 65009509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134464.2 NP_001127936.1 1 - 2 216 104
XM_006241425.1 XP_006241487.1 2 - 3 308 104
GenBank mRNA BC158738.1 - 1 - 2 218 104

Gene Symbol
LOC100911956
Entrez Gene ID
100911956
Gene Name
protein LLP homolog
Gene Aliases
-
Chromosome Location
Chr.6: 125863988 - 125865098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764900.1 XP_008763122.1 1 - 1 250 104

View Full Product Details