Assay Design



Gene Transcripts

Gene Symbol
Eid3
Entrez Gene ID
691688
Gene Name
EP300 interacting inhibitor of differentiation 3
Gene Aliases
NS4EB
Chromosome Location
Chr.7: 26976154 - 26977479 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044304.1 NP_001037769.2 1 - 1 1065 80
GenBank mRNA BC097404.1 - 1 - 1 1065 80

View Full Product Details