Assay Design
Gene Transcripts
- Gene Symbol
- Kcnc2
- Entrez Gene ID
-
246153
- Gene Name
- potassium channel, voltage gated Shaw-related subfamily C, member 2
- Gene Aliases
-
KShIIIA, Kv3.2
- Chromosome Location
-
Chr.7: 54978786 - 55159362 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M84202.1 |
- |
4 - 5 |
1866 |
140 |
X62839.1 |
- |
4 - 5 |
1963 |
140 |
View Full Product Details