Assay Design



Gene Transcripts

Gene Symbol
Best3
Entrez Gene ID
314847
Gene Name
bestrophin 3
Gene Aliases
Vmd2l3
Chromosome Location
Chr.7: 59971145 - 60057162 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191783.1 NP_001178712.1 1 - 2 143 110
XM_008765394.1 XP_008763616.1 3 - 4 1287 110

View Full Product Details