Assay Design



Gene Transcripts

Gene Symbol
Best3
Entrez Gene ID
314847
Gene Name
bestrophin 3
Gene Aliases
Vmd2l3
Chromosome Location
Chr.7: 59971145 - 60057162 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191783.1 NP_001178712.1 2 - 3 249 157
XM_008765394.1 XP_008763616.1 4 - 5 1393 157

View Full Product Details