Assay Design
Gene Transcripts
- Gene Symbol
- Trim3
- Entrez Gene ID
-
83616
- Gene Name
- tripartite motif-containing 3
- Gene Aliases
-
Berp, Rnf22
- Chromosome Location
-
Chr.1: 170441662 - 170464341 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031786.1 |
NP_113974.1 |
1 - 2 |
192 |
93 |
GenBank mRNA |
AF036255.1 |
- |
1 - 2 |
192 |
93 |
View Full Product Details