Assay Design



Gene Transcripts

Gene Symbol
Tbc1d10b
Entrez Gene ID
365372
Gene Name
TBC1 domain family, member 10b
Gene Aliases
RGD1309191
Chromosome Location
Chr.1: 198641018 - 198652674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108921.1 NP_001102391.1 8 - 9 1801 71

View Full Product Details