Assay Design



Gene Transcripts

Gene Symbol
Eif3c
Entrez Gene ID
293484
Gene Name
eukaryotic translation initiation factor 3, subunit C
Gene Aliases
Eif3s8
Chromosome Location
Chr.1: 197964406 - 197982917 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100662.1 NP_001094132.1 5 - 6 700 145
XM_006230226.2 XP_006230288.1 5 - 6 1016 145
GenBank mRNA BC169011.1 - 5 - 6 700 145
FQ225044.1 - 5 - 6 803 145
FQ227337.1 - 5 - 6 802 145

View Full Product Details