Assay Design



Gene Transcripts

Gene Symbol
Hirip3
Entrez Gene ID
361650
Gene Name
HIRA interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.1: 198298138 - 198301164 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025725.1 NP_001020896.1 6 - 7 1705 60
XM_006230307.1 XP_006230369.1 5 - 6 1402 60
GenBank mRNA BC099091.1 - 6 - 7 1705 60

View Full Product Details