Assay Design



Gene Transcripts

Gene Symbol
Enpp4
Entrez Gene ID
301261
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4
Gene Aliases
-
Chromosome Location
Chr.9: 19446893 - 19457946 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106892.1 NP_001100362.1 1 - 2 197 118

View Full Product Details