Assay Design



Gene Transcripts

Gene Symbol
Lrrc73
Entrez Gene ID
501101
Gene Name
leucine rich repeat containing 73
Gene Aliases
nod3l
Chromosome Location
Chr.9: 17111176 - 17115276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024360.2 NP_001019531.1 2 - 3 529 100
GenBank mRNA AB210855.1 - 2 - 3 429 100
BC128712.1 - 2 - 3 529 100

View Full Product Details