Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106898.1 | NP_001100368.1 | 30 - 31 | 4593 | 134 |
XM_008766928.1 | XP_008765150.1 | 14 - 15 | 2284 | 134 | |
XM_008766929.1 | XP_008765151.1 | 15 - 16 | 2177 | 134 | |
XM_008766930.1 | XP_008765152.1 | 15 - 16 | 3598 | 134 | |
XM_008766931.1 | XP_008765153.1 | 15 - 16 | 2414 | 134 | |
XM_008766932.1 | XP_008765154.1 | 13 - 14 | 2185 | 134 |