Assay Design



Gene Transcripts

Gene Symbol
Cenpq
Entrez Gene ID
363198
Gene Name
centromere protein Q
Gene Aliases
-
Chromosome Location
Chr.9: 23352271 - 23368774 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014215.1 NP_001014237.1 1 - 2 149 96
GenBank mRNA BC082113.1 - 1 - 2 149 96

View Full Product Details