Assay Design



Gene Transcripts

Gene Symbol
Smap1
Entrez Gene ID
684800
Gene Name
small ArfGAP 1
Gene Aliases
-
Chromosome Location
Chr.9: 30068674 - 30161760 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750667.3 XP_003750715.3 7 - 8 939 135
XM_003750668.3 XP_003750716.3 8 - 9 1021 135

View Full Product Details