Assay Design



Gene Transcripts

Gene Symbol
Tfap2d
Entrez Gene ID
301284
Gene Name
transcription factor AP-2 delta
Gene Aliases
Tcfap2d
Chromosome Location
Chr.9: 25304876 - 25365151 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106895.1 NP_001100365.1 3 - 4 491 65

View Full Product Details