Assay Design



Gene Transcripts

Gene Symbol
Guca1a
Entrez Gene ID
301233
Gene Name
guanylate cyclase activator 1a (retina)
Gene Aliases
-
Chromosome Location
Chr.9: 15609804 - 15620030 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106887.1 NP_001100357.1 4 - 5 724 126
XM_006244428.2 XP_006244490.1 3 - 4 679 126

View Full Product Details