Assay Design



Gene Transcripts

Gene Symbol
Dll4
Entrez Gene ID
311332
Gene Name
delta-like 4 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.3: 111135011 - 111146746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107760.1 NP_001101230.1 9 - 10 2258 131
XM_006234765.2 XP_006234827.1 9 - 10 2269 131

View Full Product Details