Assay Design



Gene Transcripts

Gene Symbol
Prnd
Entrez Gene ID
113910
Gene Name
prion protein 2 (dublet)
Gene Aliases
-
Chromosome Location
Chr.3: 124543240 - 124548548 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235056.2 XP_006235118.1 1 - 2 147 63
XM_006235057.2 XP_006235119.1 1 - 2 147 63

View Full Product Details