Assay Design
Gene Transcripts
- Gene Symbol
- Tmem62
- Entrez Gene ID
-
311350
- Gene Name
- transmembrane protein 62
- Gene Aliases
-
RGD1305647
- Chromosome Location
-
Chr.3: 112915406 - 112950418 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_230497.8 |
XP_230497.4 |
1 - 2 |
987 |
80 |
View Full Product Details