Assay Design



Gene Transcripts

Gene Symbol
Lrrc4c
Entrez Gene ID
311236
Gene Name
leucine rich repeat containing 4C
Gene Aliases
RGD1311013
Chromosome Location
Chr.3: 85421169 - 85994934 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107753.1 NP_001101223.1 - 1353 152

View Full Product Details