Assay Design



Gene Transcripts

Gene Symbol
Shf
Entrez Gene ID
362205
Gene Name
Src homology 2 domain containing F
Gene Aliases
RGD1562705
Chromosome Location
Chr.3: 114288021 - 114307328 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234884.2 XP_006234946.1 2 - 3 337 70

View Full Product Details